4-(Dimethylamino)phenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09011-04
L09011-09
AAAL09011-04EA
71.3
USD
AAAL09011-04
AAAL09011-09
4-(Dimethylamino)phenyl isothiocyanate 97%
4-(Dimethylamino)phenyl isothiocyanate
Purity: 97%
CAS No.: 2131-64-8
Molecular Formula: C9H10N2S
Formula Weight: 178.25
Storage Temperature: 2°C to 8°C
Odor: Irritating
Boiling Point: 152°/6mm
Beilstein Reference: 388270
UN No.: 2928
MDL No.: MFCD00041093
Formula:
C₉H₁₀N₂S Boiling Pt: 152 °C/6 mm Melting Pt: 64…70 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00041093 CAS Number: 2131-64-8 EINECS: 218-360-4 UN: 2928 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...