2-Hydrazinoquinoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Quinolylhydrazine, 2-Hydrazinylquinoline
H50700-03
H50700-MD
AAAH50700-03EA
251.71
USD
AAAH50700-03
AAAH50700-MD
2-Hydrazinoquinoline 97%
2-Hydrazinoquinoline
Purity: 97%
CAS No.: 15793-77-8
Molecular Formula: C9H9N3
Formula Weight: 159.19
Melting Point: 141-143°
MDL No.: MFCD00041860
Formula:
C₉H₉N₃ MW: 159.19 g/mol Melting Pt: 141…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041860 CAS Number: 15793-77-8 EINECS: 239-899-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...