3-(3-Bromophenyl)-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(3-Bromophenyl)-1H-pyrazole

H50438-06 H50438-03 H50438-03 H50438-06
AAAH50438-06EA 542.99 USD
AAAH50438-06 CAAAAH50438-06 AAAH50438-03 CAAAAH50438-03
3-(3-Bromophenyl)-1H-pyrazole 98%
3-(3-Bromophenyl)-1H-pyrazole
Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01940433
CAS Number: 149739-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR