Diethyl (2-oxopropyl)phosphonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyl acetylmethylphosphonate
H26896-06
H26896-14
AAH26896-06EA
51.5
USD
AAH26896-06
AAH26896-03
AAH26896-14
Diethyl (2-oxopropyl)phosphonate 97%
Diethyl (2-oxopropyl)phosphonate
Purity: 97%
CAS No.: 1067-71-6
Molecular Formula: C7H15O4P
Formula Weight: 194.17
Flash Point: 113°(235°F)
Refractive Index: 1.433
Boiling Point: 129°/9mm
MDL No.: MFCD00044728
Formula:
C₇H₁₅O₄P Boiling Pt: 129 °C/9 mm Density: 1.01 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044728 CAS Number: 1067-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...