1H,1H,2H,2H-Perfluorooctyltriethoxysilane 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;

B24620-06 B24620-14
AAB24620-06EA 125.82 USD
AAB24620-06 AAB24620-14
1H,1H,2H,2H-Perfluorooctyltriethoxysilane 97%
1H,1H,2H,2H-Perfluorooctyltriethoxysilane

Purity: 97% CAS No.: 51851-37-7 Molecular Formula: C14H19F13O3Si Formula Weight: 510.37 Flash Point: 97°(206°F) Refractive Index: 1.345 Melting Point: -38° Boiling Point: 220° MDL No.: MFCD00042333

Formula: C₁₄H₁₉F₁₃O₃Si
MW: 510.367 g/mol
Boiling Pt: 220 °C
Melting Pt: <–38 °C
Density: 1.333
Flash Pt: 97 °C(206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042333
CAS Number: 51851-37-7
EINECS: 257-473-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR