Benzo[b]furan-2-boronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid

B23676-03 B23676-06 B23676-14
AAB23676-03EA 51.2 USD
AAB23676-03 AAB23676-06 AAB23676-14
Benzo[b]furan-2-boronic acid 98%
Benzo[b]furan-2-boronic acid

Purity: 98% CAS No.: 98437-24-2 Molecular Formula: C8H7BO3 Formula Weight: 161.95 Storage Temperature: 2°C to 8°C

Formula: C₈H₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD00236019
CAS Number: 98437-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR