(+/-)-ɑ-Bisabolol 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

B21457-18 B21457-09 B21457-18 B21457-09
AAB21457-18EA 225 USD
AAB21457-18 AAB21457-09 CAAAB21457-09 CAAAB21457-18
(+/-)-ɑ-Bisabolol 96%
(+/-)-alpha-Bisabolol
Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Density: 0.93
Flash Pt: 135 °C(275 °F)
Storage Temperature: Ambient
MDL Number: MFCD03846910
CAS Number: 515-69-5
EINECS: 208-205-9
Merck Index: 14,1241

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR