6-Aminophthalide 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Aminoisobenzofuran-1(3H)-one, 6-AMINOPHTHALIDE

A18896-88 A18896-09 A18896-02
AAA18896-88EA 243 USD
AAA18896-88 AAA18896-09 AAA18896-02
6-Aminophthalide 95%
6-Aminophthalide

Purity: 95% CAS No.: 57319-65-0 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Melting Point: 182-185° MDL No.: MFCD00033530

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 182…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00033530
CAS Number: 57319-65-0
EINECS: 260-675-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR