6-Amino-2-methylquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-methylquinolin-6-amine

A13967-06 A13967-03 A13967-MD
AAA13967-06EA 591 USD
AAA13967-06 AAA13967-03 AAA13967-MD
6-Amino-2-methylquinoline 97%
6-Amino-2-methylquinoline

Purity: 97% CAS No.: 65079-19-8 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 189-191° Beilstein Reference: 114795

Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00052600
CAS Number: 65079-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR