2,4,6-Trimethoxyboroxin
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethoxycyclotriboroxane, Trimethoxyboroxine
089153-14
089153-30
AA89153-14EA
27.2
USD
AA89153-14
AAA89153-18
AAA89153-30
2,4,6-Trimethoxyboroxin
2,4,6-Trimethoxyboroxin
Total boroxine, 100%; hydroxide content,
CAS No.: 102-24-9
Molecular Formula: (CH3O)3B3O3
Formula Weight: 173.53
Flash Point: 14°(57°F)
Physical Form: Liquid
Density: 1.216
Refractive Index: 1.3996
Sensitivity: Air and Moisture Sensitive
Odor: Strong
Melting Point: 10-11°
Boiling Point: 130°
Beilstein Reference: 1709158
UN No.: 2924
MDL No.: MFCD00005946
Formula:
C₃H₉B₃O₆ Boiling Pt: 130 °C Melting Pt: 10…11 °C Density: 1,216 Flash Pt: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005946 CAS Number: 102-24-9 EINECS: 203-016-8 UN: 2924 ADR: 3,II |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...