Titanium nickel composite (70:30), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

088384-36 088384-22 088384-09
AA88384-36EA 1246 USD
AA88384-36 AA88384-22 AA88384-09
Titanium nickel composite (70:30), powder -325 mesh
Titanium nickel composite

Molecular Formula: Ti:Ni; 70:30 wt% Sensitivity: Air Sensitive UN No.: 3089 MDL No.: MFCD02091734

Formula: Ti:Ni; 50:50 %
Storage Temperature: Ambient
MDL Number: MFCD02091734
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR