Aluminium oxide neutral HPLC flash grade

Supplier: Thermo Scientific Chemicals

045985-30 045985-A1
AA45985-A1EA 241 USD
AA45985-A1 AA45985-30
Aluminium oxide neutral HPLC flash grade
Aluminium oxide neutral
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR