Silica gel, Pore size: 120 Å ≥99.99%, spherical 5 µm APS for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
045810-06
045810-14
AA45810-06EA
133
USD
AA45810-06
AA45810-14
Silica gel, Pore size: 120 Å ≥99.99%, spherical 5 µm APS for HPLC
Silica gel
Purity: 99.99+%
CAS No.: 63231-67-4
Physical Form: S.A. 340m2/g, P.V. 1.00cc/g
MDL No.: MFCD00011232
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...