Silica gel, Pore size: 70 Å ≥99.99%, spherical 5 µm APS for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
045809-06
045809-14
AA45809-06EA
130
USD
AA45809-06
AA45809-14
Silica gel, Pore size: 70 Å ≥99.99%, spherical 5 µm APS for HPLC
Silica gel
Purity: 99.99+%
CAS No.: 63231-67-4
Physical Form: S.A. 500m2/g, P.V. 0.95cc/g
Odor: Odorless
pH: 5.5-8
MDL No.: MFCD00011232
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...