Palladium(II) nitrate (4 - 5% Pd) (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

045512-NB 045512-ND
AA45512-NBEA 1163 USD
AA45512-NB AA45512-ND
Palladium(II) nitrate (4 - 5% Pd) (w/w) in aqueous solution
Palladium (II) nitrate
Formula: N₂O₆Pd
MW: 230.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011169
CAS Number: 10102-05-3
EINECS: 233-265-8
UN: 3264
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR