Polyethyleneimine, M.W. M.W. 50,000 - 100,000 30% (w/w) in aqueous solution, branched

Supplier: Thermo Scientific Chemicals
Warning

045024-14 045024-22 045024-36
AA45024-14EA 66.97 USD
AA45024-14 AA45024-22 AA45024-36
Polyethyleneimine, M.W. M.W. 50,000 - 100,000 30% (w/w) in aqueous solution, branched
Polyethyleneimine

Polyethyleneimine acts as a protein precipitant used to purify proteins. It is used as a chelating agent and as a scavenger for aldehydes and oxides. It is also used in detergents, paper industry, dyes, printing inks and in water treatment.


CAS No.: 9002-98-6 Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y Physical Form: Liquid Appearance: Colorless to pale yellow Odor: Odorless MDL No.: MFCD00803910

Formula: (CH2CH2NH)n
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR