Nickel etchant for evaporated nickel films
Supplier: Thermo Scientific Chemicals
|
Danger
|
044586-LV
044586-R0
AA44586-LVEA
195.99
USD
AA44586-LV
AA44586-R0
Nickel etchant for evaporated nickel films
Nickel etchant
Applications: Standard nickel etchant for evaporated nickel films
Storage Temperature:
Ambient |
MDL Number:
MFCD09039236 UN: 3264 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...