Nickel etchant for electrodeposited nickel films
Supplier: Thermo Scientific Chemicals
|
Danger
|
044580-R0
AA44580-R0EA
94.59
USD
AA44580-R0
Nickel etchant for electrodeposited nickel films
Nickel etchant
Applications: High purity etchant for electrodeposited nickel films
Storage Temperature:
Ambient |
MDL Number:
MFCD09039236 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...