Palladium 10% on activated carbon powder standard wetted with ca. 50% water, reduced Type 58
Supplier: Thermo Scientific Chemicals
44350-22
44350-06
44350-14
44350-22
44350-14
44350-06
AA44350-22EA
2783.99
USD
AA44350-22
CAAA44350-22
AA44350-06
CAAA44350-06
AA44350-14
CAAA44350-14
Palladium 10% on activated carbon powder standard wetted with ca. 50% water, reduced Type 58
Palladium
Applications: Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic nitrites to 1° amines
Formula:
Pd MW: 106.420 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03457879 CAS Number: 7440-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...