Silver, Oil Based Standard Solution, Specpure®, Ag 5000 µg/g, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
43868-18
AA43868-18EA 72.08 USD
AA43868-18
Silver, Oil Based Standard Solution, Specpure®, Ag 5000 µg/g, Thermo Scientific
Standards Cationic Standards Silver Standards
Physical Form: Liquid
MDL No.: MFCD00003397
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR