Coal tar solution contains 81.0-86.0% alcohol
Supplier: Thermo Scientific Chemicals
|
Danger
|
043671-AP
043671-K4
AA43671-APEA
56.45
USD
AA43671-AP
AA43671-K4
Coal tar solution contains 81.0-86.0% alcohol
Coal tar
CAS No.: 8007-45-2
Flash Point: 13°(55°F)
Physical Form: Liquid
Appearance: Dark brown
Odor: Alcohol-like
UN No.: 1993
MDL No.: MFCD00146482
Flash Pt:
13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00146482 CAS Number: 8007-45-2 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...