Gold zinc alloy (90:10 w%) ≥99.95% (metals basis), slug 3.175mm (0.125in) dia x 3.175mm (0.125in) length
Supplier: Thermo Scientific Chemicals
043470-KG
043470-KJ
AA43470-KGEA
180
USD
AA43470-KG
AA43470-KJ
Gold zinc alloy (90:10 w%) ≥99.95% (metals basis), slug 3.175mm (0.125in) dia x 3.175mm (0.125in) length
Gold Zinc alloy (90:10 w%)
Purity: 99.95%
Molecular Formula: Au:Zn; 90:10 wt%
Physical Form: ≈0.45g/slug
MDL No.: MFCD02092916
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...