Ruthenium(IV) sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

043098-06 43098-06
AA43098-06EA 769 USD
AA43098-06 CAAA43098-06
Ruthenium(IV) sulfide ≥99.9% (metals basis)
Ruthenium(IV) sulfide
Formula: RuS₂
MW: 165.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799846
CAS Number: 12166-20-0
EINECS: 235-318-0
UN: 3178
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR