Ground & Waste Water Pollution Standard Solution 5, Specpure®

Supplier: Thermo Scientific
Specpure®
42580-AE 042580-AE 42580-AP 42580-AP
AA42580-AEEA 133 USD
AA42580-AE CAAA42580-AE AA42580-AP CAAA42580-AP
Ground & Waste Water Pollution Standard Solution 5, Specpure®
Standards Environmental Analysis Standards
For US EPA Clean Water Act.

  • 4 Analytes

UN No.: 3264
MDL No.: MFCD03792307
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR