Chromium, Oil based standard solution, Specpure®, Cr 5000 µg/g, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
42438-18
AA42438-18EA 81.52 USD
AA42438-18
Chromium, Oil based standard solution, Specpure®, Cr 5000 µg/g, Thermo Scientific
Standards Cationic Standards Chromium Standards
Physical Form: Liquid
MDL No.: MFCD00010944
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR