Aluminum, Oil based standard solution, Specpure®, Al 5000 µg/g, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
42432-18
AA42432-18EA 75.6 USD
AA42432-18
Aluminum, Oil based standard solution, Specpure®, Al 5000 µg/g, Thermo Scientific
Standards Cationic Standards Aluminum Standards
Physical Form: Liquid
MDL No.: MFCD00134029
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR