Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness 0.2 mm (0.008 in) Type 304
Supplier: Thermo Scientific Chemicals
|
Danger
|
042255-FI
042255-GH
AA42255-FFEA
27.44
USD
AA42255-FF
AA42255-FI
AA42255-GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness 0.2 mm (0.008 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odor: Odorless
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...