Dysprosium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dysprosium(III) perchlorate

040574-NA 40574-NA
AA40574-NAEA 64.43 USD
AA40574-NA CAAA40574-NA
Dysprosium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade
Dysprosium (III) perchlorate hexahydrate
Formula: Cl₃DyO₁₂·6H₂O
MW: 568.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150227
CAS Number: 14692-17-2
EINECS: 237-838-3
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR