Cadmium molybdenum oxide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

040202-30 40202-30
AA40202-30EA 534 USD
CAAA40202-30 AA40202-30
Cadmium molybdenum oxide ≥99.5% (metals basis)
Cadmium molybdenum oxide

Slightly soluble in water. Soluble in acid


Applications: Electronic and optical applications

Formula: CdMoO₄
MW: 272.35 g/mol
Melting Pt: ∼ 1250 °C
Storage Temperature: Ambient
MDL Number: MFCD00015997
CAS Number: 13972-68-4
EINECS: 237-752-6
UN: 2570
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR