Thallium(I) hexafluorophosphate ≥97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Thallium(I) hexafluorophosphate(V)
039627-06
39627-06
AA39627-06EA
258
USD
CAAA39627-06
AA39627-06
Thallium(I) hexafluorophosphate ≥97%
Thallium (I) hexafluorophosphate
Formula:
F₆PTl Storage Temperature: Ambient |
MDL Number:
MFCD00049807 CAS Number: 60969-19-9 UN: 1707 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...