Multi-Element QC-19 Check Standard Solution, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

039146-AP 039146-AE
AA39146-APEA 852 USD
AA39146-AP AA39146-AE
Multi-Element QC-19 Check Standard Solution, Specpure®, Thermo Scientific
Standards Cationic Standards Multi-Element Standards

Molecular Formula: Matrix: 5% HNO3/tr. F-/tr. tartaric acid
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00210655

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR