Carnauba wax

Supplier: Thermo Scientific Chemicals

031117-A1 031117-A7 031117-30
AA31117-A1EA 108.99 USD
AA31117-A1 AA31117-A7 AA31117-30
Carnauba wax
Carnauba wax

CAS No.: 8015-86-9 Physical Form: Flakes Density: 0.990-0.999 Refractive Index: 1.4500 Odor: Odorless Melting Point: 82-86° MDL No.: MFCD00130724

Melting Pt: 82…86 °C
Density: 0.990…0.999
Storage Temperature: Ambient
MDL Number: MFCD00130724
CAS Number: 8015-86-9
EINECS: 232-399-4
Merck Index: 14,1846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR