Vinylferrocene
Supplier: Thermo Scientific Chemicals
17450-03
17450.06
17450.03
17450-06
AA17450-03EA
120.99
USD
AA17450-03
CAAA17450-03
AA17450-06
CAAA17450-06
Vinylferrocene
Vinylferrocene
Formula:
C₁₂H₁₂Fe Boiling Pt: 80…85 °C (0.7 mmHg) Melting Pt: 52…55 °C Flash Pt: 62 °C (144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001434 EINECS: 215-041-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...