Lithium, Oil Based Standard Solution, Specpure®, Li 1000 µg/g, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
14215-18
AA14215-18EA 73.49 USD
AA14215-18
Lithium, Oil Based Standard Solution, Specpure®, Li 1000 µg/g, Thermo Scientific
Standards Cationic Standards Lithium Standards
Physical Form: Liquid
MDL No.: MFCD00134051
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR