Sodium, Oil based standard solution, Specpure®, Na 1000 µg/g, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
14212-18
AA14212-18EA 68.47 USD
AA14212-18
Sodium, Oil based standard solution, Specpure®, Na 1000 µg/g, Thermo Scientific
Standards Cationic Standards Sodium Standards
Physical Form: Liquid
MDL No.: MFCD00085307
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR