Lead, Oil Based Standard Solution, Specpure®, Pb 1000 µg/g, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
14208-18
AA14208-18EA 72.17 USD
AA14208-18
Lead, Oil Based Standard Solution, Specpure®, Pb 1000 µg/g, Thermo Scientific
Standards Cationic Standards Lead Standards
Physical Form: Liquid
MDL No.: MFCD00134050
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR