Cerium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cerium(III) chloride hydrateCerous chloride

011325-18 11325-30 011325-30 11325-18
AA11325-30EA 169.99 USD
CAAA11325-30 AA11325-30 CAAA11325-18 AA11325-18
Cerium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)
Cerium trichloride hydrate

Cerium(III) chloride hydrate is used as an important raw material the preparation of other cerium salts such as cerium(III) trifluoromethanesulfonate. It is used in the conversion of esters to allylsilanes.


Sensitivity: Air Sensitive and Hygroscopic
Appearance: White crystalline aggregates

Formula: CeCl₃·7H₂O
MW: 246.48 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149633
CAS Number: 19423-76-8
EINECS: 232-227-8
Merck Index: 12,02041

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR