OxyDish™ Anaerobic Plate, Oxyrase®
Supplier: Oxyrase
A-OD-0100
101975-862BX 150.89 USD
101975-862
OxyDish™ Anaerobic Plate, Oxyrase®
Media for Microbiology Prepared Media for Microbiology
The patented OxyDish™ is specially designed to create a seal that maintains anaerobiosis, after Oxyrase® for Agar (OA) has been added to a poured agar medium.
The OxyDish™ is very similar to a standard culture dish. However, there are some very significant differences of note. The base is the same size as a regular Petri dish, but contains an isomark useful for automation purposes. The lid contains a sealing ring to set over the dish base after it has been filled. When the base is filled with solidified agar and the dish inverted, the agar surface will come to rest on this sealing ring.
Ordering information: Each individually gamma irradiated sleeve contains 20 sterile OxyDish™ plates.