Iodosobenzene diacetate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Iodobenzene diacetate, (Diacetoxyiodo)benzene

B24531-14 B24531-22 B24531-36
AAB24531-14EA 102.13 USD
AAB24531-14 AAB24531-22 AAB24531-36
Iodosobenzene diacetate 98+%
Iodosobenzene diacetate

Fieser: 1,508 15,258 16,275 17,280 18,290 19,268 20,305 21,343


Purity: 98+% CAS No.: 3240-34-4 Molecular Formula: C10H11IO4 Formula Weight: 322.10 Melting Point: 159-161° Beilstein Reference: 1879369

Formula: C₁₀H₁₁IO₄
MW: 322.1 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00008692
CAS Number: 3240-34-4
EINECS: 221-808-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR