Zinc selenide
Supplier: Thermo Scientific Chemicals
|
Danger
|
A19947-09
A19947-18
A19947-30
AAA19947-09EA
51.4
USD
AAA19947-09
AAA19947-18
AAA19947-30
Zinc selenide
Zinc selenide
CAS No.: 1315-09-9
Molecular Formula: ZnSe
Formula Weight: 144.33
Odor: Odorless
Melting Point:
Formula:
SeZn Melting Pt: >1100 °C Density: 5.42^1^5 Storage Temperature: Ambient |
MDL Number:
MFCD00011296 CAS Number: 1315-09-9 EINECS: 215-259-7 UN: 3283 ADR: 6.1,III Merck Index: 14,10156 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...