Aluminium oxide activated basic HPLC flash grade
Supplier: Thermo Scientific Chemicals
045949-30
045949-A1
AA45949-30EA
70
USD
AA45949-30
AA45949-A1
Aluminium oxide activated basic HPLC flash grade
Aluminium oxide activated basic
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 18-32 Micron APS Powder
Refractive Index: 1.765
Appearance: White
Odor: Odorless
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Formula:
Al₂O₃ Boiling Pt: 2980 °C Melting Pt: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...