Antimony(III) telluride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
045922-09
045922-18
AA45922-09EA
167
USD
AA45922-09
AA45922-18
Antimony(III) telluride ≥99.999% (metals basis)
Antimony (III) telluride
Purity: 99.999%
CAS No.: 1327-50-0
Molecular Formula: Sb2Te3
Formula Weight: 626.30
Physical Form: -100 Mesh Powder
Density: 6.5
Appearance: Grey
Odor: Odorless
Melting Point: 629°
UN No.: 1549
MDL No.: MFCD00016320
Formula:
Sb₂Te₃ MW: 626.32 g/mol Melting Pt: 620 Density: 6.5 Storage Temperature: Ambient |
MDL Number:
MFCD00016320 CAS Number: 1327-50-0 EINECS: 215-480-9 UN: 1549 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...