Silver chromate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Silver(I) chromate
, Argentous chromate
045495-22
045495-06
AA45495-06EA
30.6
USD
AA45495-06
AA45495-22
Silver chromate ≥99.9% (metals basis)
Silver chromate
Formula:
Ag₂CrO₄ MW: 331.73 g/mol Boiling Pt: 1550 °C (1013 hPa) Density: 5.63 Storage Temperature: Ambient |
MDL Number:
MFCD00003402 CAS Number: 7784-01-2 EINECS: 232-043-8 UN: 1479 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...