Anti-IL-1b Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: H1b-98]

Supplier: Biolegend
511709 511710
511709-BLEA 158.8 USD
511709-BL 511710-BL
Anti-IL-1b Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: H1b-98]
Antibodies
Pacific Blue™ anti-human IL-1β [H1b-98]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

* Pacific Blue™ has a maximum emission of 455 nm when it is excited at 405 nm. Prior to using Pacific Blue™ conjugate for flow cytometric analysis, please verify your flow cytometer's capability of exciting and detecting the fluorochrome.

Type: Primary
Antigen: IL-1b
Clonality: Monoclonal
Clone: H1b-98
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR