Anti-IgG1 Mouse Antibody [clone: IL2R.1]

Supplier: Diagnostic BioSystems

PDM144 PDM144
95029-762EA 301.83 USD
95029-762 CA95029-762
Anti-IgG1 Mouse Antibody [clone: IL2R.1]
Antibodies
Type:
Antigen: IL2RA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR