WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
07-0020B-1000 07-0020B-101 07-0020B-1200 07-0020B-1202 07-0020B-201 07-0020B-2200 07-0020B-300 07-0020G-1000 07-0020G-101 07-0020G-1200 07-0020G-1202 07-0020G-201 07-0020G-2200 07-0020G-300 07-0020N-1000 07-0020N-101 07-0020N-1200 07-0020N-1202 07-0020N-201 07-0020N-2200 07-0020N-300 07-0020R-1000 07-0020R-101 07-0020R-1200 07-0020R-1202 07-0020R-201 07-0020R-2200 07-0020R-300 07-0021B-1000 07-0021B-101 07-0021B-1200 07-0021B-1202 07-0021B-201 07-0021B-2200 07-0021B-300 07-0021G-1000 07-0021G-101 07-0021G-1200 07-0021G-1202 07-0021G-201 07-0021G-2200 07-0021G-300 07-0021N-1000 07-0021N-101 07-0021N-1200 07-0021N-1202 07-0021N-201 07-0021N-2200 07-0021N-300 07-0021R-1000 07-0021R-101 07-0021R-1200 07-0021R-1202 07-0021R-201 07-0021R-2200 07-0021R-300 07-0045B-1000 07-0045B-101 07-0045B-1200 07-0045B-1202 07-0045B-201 07-0045B-201-S 07-0045B-2200 07-0045B-300 07-0045G-1000 07-0045G-101 07-0045G-1200 07-0045G-1202 07-0045G-201 07-0045G-2200 07-0045G-300 07-0045N-1000 07-0045N-101 07-0045N-1200 07-0045N-1202 07-0045N-201 07-0045N-2200 07-0045N-300 07-0045R-1000 07-0045R-101 07-0045R-1200 07-0045R-1202 07-0045R-201 07-0045R-2200 07-0045R-300 07-0075B-1000 07-0075B-1202 07-0075B-201 07-0075B-2200 07-0075B-300 07-0075G-1000 07-0075G-101 07-0075G-1200 07-0075G-1202 07-0075G-201 07-0075G-2200 07-0075G-300 07-0075N-1000 07-0075N-101 07-0075N-1200 07-0075N-1202 07-0075N-201 07-0075N-2200 07-0075N-300 07-0075R-1000 07-0075R-101 07-0075R-1200 07-0075R-1202 07-0075R-201 07-0075R-2200 07-0075R-300 07-0070B-1000 07-0070B-101 07-0070B-1200 07-0070B-1202 07-0070B-201 07-0070B-2200 07-0070B-300 07-0070G-1000 07-0070G-101 07-0070G-1200 07-0070G-1202 07-0070G-201 07-0070G-2200 07-0070G-300 07-0070N-1000 07-0070N-101 07-0070N-1200 07-0070N-1202 07-0070N-201 07-0070N-2200 07-0070N-300 07-0070R-1000 07-0070R-101 07-0070R-1200 07-0070R-1202 07-0070R-201 07-0070R-300 07-0020B-250 07-0020B-350 07-0020G-250 07-0020G-350 07-0020N-250 07-0020N-350 07-0020R-250 07-0020R-350 07-0021B-250 07-0021B-350 07-0021G-250 07-0021G-350 07-0021N-250 07-0021N-350 07-0021R-250 07-0021R-350 07-0045B-250 07-0045B-350 07-0045G-250 07-0045G-350 07-0045N-250 07-0045N-350 07-0045R-250 07-0045R-350 07-0070B-250 07-0070B-350 07-0070G-250 07-0070G-350 07-0070N-250 07-0070N-350 07-0070R-250 07-0070R-350 07-0075B-250 07-0075B-350 07-0075G-250 07-0075G-350 07-0075N-250 07-0075N-350 07-0075R-250 07-0075R-350
97044-572PK 141.18 USD
97044-572 97044-576 97044-578 97044-580 97044-582 97044-584 97044-586 97044-590 97044-594 97044-596 97044-598 97044-600 97044-602 97044-604 97044-616 97044-620 97044-622 97044-624 97044-626 97044-628 97044-630 97044-634 97044-638 97044-640 97044-642 97044-644 97044-646 97044-648 97044-652 97044-656 97044-658 97044-660 97044-662 97044-664 97044-666 97044-670 97044-674 97044-676 97044-678 97044-680 97044-682 97044-684 97044-696 97044-700 97044-702 97044-704 97044-706 97044-708 97044-710 97044-714 97044-718 97044-720 97044-722 97044-724 97044-726 97044-728 97044-742 97044-746 97044-748 97044-750 97044-752 97044-754 97044-756 97044-758 97044-762 97044-766 97044-768 97044-770 97044-772 97044-774 97044-776 97044-790 97044-794 97044-796 97044-798 97044-800 97044-802 97044-804 97044-808 97044-812 97044-814 97044-816 97044-818 97044-820 97044-822 97044-838 97044-842 97044-846 97044-848 97044-850 97044-852 97044-856 97044-860 97044-862 97044-864 97044-866 97044-868 97044-870 97044-882 97044-886 97044-888 97044-890 97044-892 97044-894 97044-896 97044-900 97044-904 97044-906 97044-908 97044-910 97044-912 97044-914 89231-708 89231-712 89231-714 89231-716 89231-718 89231-720 89231-722 89231-726 89231-730 89231-732 89231-734 89231-736 89231-738 89231-740 89231-744 89231-748 89231-750 89231-752 89231-754 89231-756 89231-758 89231-762 89231-766 89231-768 89231-770 89231-772 89231-776 30618-124 30618-126 30618-128 30618-130 30618-132 30618-134 30618-136 30618-138 30618-140 30618-142 30618-144 30618-146 30618-148 30618-150 30618-152 30618-154 30618-156 30618-158 30618-160 30618-162 30618-164 30618-166 30618-168 30618-170 30618-172 30618-174 30618-176 30618-178 30618-180 30618-182 30618-184 30618-186 30618-188 30618-190 30618-192 30618-194 30618-196 30618-198 30618-200 30618-202
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Microplates Deep Well Microplates
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with glass inserts and color-coded caps.
  • Variety of volumes available
  • Standard chromatography closures
  • Caps with septa
  • Silicone mats are resealable
A comprehensive line of 96-Well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials
Polypropylene μLPlates® have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. Users may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system allows even GC samples to be converted to 96-Well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/Silicone rubber using a mechanical seal. A variety of insert options allows users to choose the specifications appropriate for any application.
The system combines microplates, glass inserts, μLmats™, and caps with chromatography-based septa to offer secure vessels to maintain even the most volatile samples.