Anti-IgG Goat Polyclonal Antibody

Supplier: GeneTex
GTX77542
89353-780EA 220.67 USD
89353-780
Anti-IgG Goat Polyclonal Antibody
Antibodies
Goat anti-Human IgG (Fc fragment) polyclonal antibody

Works in most applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR