Anti-Ku80 Mouse Monoclonal Antibody [clone: 9403]

Supplier: GeneTex
GTX70274
89364-844EA 600.25 USD
89364-844
Anti-Ku80 Mouse Monoclonal Antibody [clone: 9403]
Antibodies
Mouse monoclonal antibody to human Ku80

  • For WB: Use at a dilution of 1:1000-1:5000. For ICC/IF: Use at a dilution of 1:1000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: Ku80
Clonality: Monoclonal
Clone: 9403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR