Anti-A1AT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX29373
89352-606EA 673.6 USD
89352-606
Anti-A1AT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to alpha 1 Antitrypsin

For IHC-P: Use at a dilution of 1:25-1:50 in an ABC method. An incubation of 30 minutes at RT is recommended. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: A1AT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR