Anti-PXN Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24832
89361-536EA 634.48 USD
89361-536
Anti-PXN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Paxillin ( phospho Y31 )

WB: Use at a dilution of 1/1000. Predicted molecular weight: 68 kDa. Can be blocked with GTX25248 Paxillin phospho Y31 and non-phospho peptides (pair) Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR